2-Amino-3-bromo-5-methylpyridine ≥98.0%
Supplier: TCI America
A1555-5G
A1555-25G
TCA1555-5GEA
55.3
CAD
TCA1555-5G
TCA1555-25G
2-Amino-3-bromo-5-methylpyridine ≥98.0%
2-Amino-3-bromo-5-methylpyridine
CAS Number: 17282-00-7
MDL Number: MFCD00068231
Molecular Formula: C6H7BrN2
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Other Product Names / Synonyms: 6-Amino-5-bromo-3-picoline
Formula:
C₆H₇BrN₂ Melting Pt: 72 °C |
MDL Number:
MFCD00068231 CAS Number: 17282-00-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...