2-Amino-3-bromo-5-nitrobenzonitrile ≥98.0%
Supplier: TCI America
A1553-25G
A1553-5G
TCA1553-25GEA
505.76
CAD
TCA1553-25G
TCA1553-5G
2-Amino-3-bromo-5-nitrobenzonitrile ≥98.0%
2-Amino-3-bromo-5-nitrobenzonitrile
CAS Number: 17601-94-4
MDL Number: MFCD00054185
Molecular Formula: C7H4BrN3O2
Molecular Weight: 242.03
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 191
Formula:
C₇H₄BrN₃O₂ Melting Pt: 191 °C |
MDL Number:
MFCD00054185 CAS Number: 17601-94-4 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...