4-Vinylphenyl acetate ≥98.0% stabilized

Supplier: TCI America

A1551-5G A1551-25G
TCA1551-005GEA 48.98 CAD
TCA1551-005G TCA1551-25G
4-Vinylphenyl acetate ≥98.0% stabilized
4-Vinylphenyl acetate

(stabilized with TBC)
CAS Number: 2628-16-2
MDL Number: MFCD00075734
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 87
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: 4-Acetoxystyrene, Acetic Acid 4-Vinylphenyl Ester

Formula: C₁₀H₁₀O₂
Density: 1.07 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00075734
CAS Number: 2628-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=746ebe9e3f89e96c7633efcec72d65f646bad0bc&vid=746ebe9e3f89e96c7633efcec72d65f646bad0bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR