N-Acetyl-L-phenylalanine 99,0

Supplier: TCI America

Synonyms: Ac-Phe-OH

A1541-5G A1541-25G
TCA1541-005GEA 28.95 CAD
TCA1541-005G TCA1541-25G
N-Acetyl-L-phenylalanine 99,0
N-Acetyl-L-phenylalanine

CAS Number: 2018-61-3
MDL Number: MFCD00063158
Molecular Formula: C11H13NO3
Molecular Weight: 207.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 168
Specific rotation [a]20/D: 40 deg (C=5, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Phe-OH

Formula: C11H13NO3
MW: 207.2 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00063158
CAS Number: 2018-61-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4660d750a122f7dbce1d80303a4ee9bc5392a83b&vid=4660d750a122f7dbce1d80303a4ee9bc5392a83b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR