4-Aminophthalic acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
A1512-25G
A1512-5G
TCA1512-25GEA
428.18
CAD
TCA1512-25G
TCA1512-5G
4-Aminophthalic acid ≥95.0% (by titrimetric analysis)
4-Aminophthalic acid
CAS Number: 5434-21-9
MDL Number: MFCD00013985
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 344
Formula:
C₈H₇NO₄ MW: 181.15 g/mol Melting Pt: 344 °C |
MDL Number:
MFCD00013985 CAS Number: 5434-21-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...