D-(+)-Allose ≥98.0% (by HPLC)

Supplier: TCI America

A1488-100MG A1488-1G
TCA1488-100MGEA 107.72 CAD
TCA1488-100MG TCA1488-001G
D-(+)-Allose ≥98.0% (by HPLC)
D(+)-Allose

CAS Number: 2595-97-3
MDL Number: MFCD00135833
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 149
Specific rotation [a]20/D: 14 deg (C=5, H2O)

Formula: C₆H₁₂O₆
Melting Pt: 149 °C
MDL Number: MFCD00135833
CAS Number: 2595-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0269c5af634fe781c0996dac823b974f98a7093f&vid=0269c5af634fe781c0996dac823b974f98a7093f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR