5-Amino-2-mercaptobenzimidazole ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Amino-1H-benzoimidazole-2-thiol

A1482-5G A1482-25G
TCA1482-005GEA 41.08 CAD
TCA1482-005G TCA1482-25G
5-Amino-2-mercaptobenzimidazole ≥97.0% (by titrimetric analysis)
5-Amino-2-mercaptobenzimidazole

CAS Number: 2818-66-8
MDL Number: MFCD00022671
Molecular Formula: C7H7N3S
Molecular Weight: 165.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 237


Other Product Names / Synonyms: 5-Amino-2-benzimidazolethiol

Formula: C₇H₇N₃S
MW: 165.22 g/mol
Melting Pt: 237 °C
MDL Number: MFCD00022671
CAS Number: 2818-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b58ffe23f52cb1c4b398518f1febfce927f6b67&vid=7b58ffe23f52cb1c4b398518f1febfce927f6b67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR