Allylpalladium(II) chloride dimer ≥97.0%
Supplier: TCI America
Synonyms:
Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer
CAS Number: 12012-95-2
MDL Number: MFCD00044874
Molecular Formula: C6H10Cl2Pd2
Molecular Weight: 365.89
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Di-mu-chlorobis[(eta-allyl)palladium(II)]
Formula:
C₆H₁₀Cl₂Pd₂ MW: 365.89 g/mol |
MDL Number:
MFCD00044874 CAS Number: 12012-95-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...