Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97.0%
Supplier: TCI America
A1475-5G
A1475-25G
TCA1475-005GEA
39.5
CAD
TCA1475-005G
TCA1475-25G
Ethyl-3-amino-4,4,4-trifluorocrotonate ≥97.0%
Ethyl 3-amino-4,4,4-trifluorocrotonate
CAS Number: 372-29-2
MDL Number: MFCD00068195
Molecular Formula: C6H8F3NO2
Molecular Weight: 183.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 83
Flash Point (°C): 65
Freezing point (°C): 24
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-4,4,4-trifluorocrotonic Acid Ethyl Ester
Formula:
C₆H₈F₃NO₂ Boiling Pt: 83 °C Flash Pt: 65 °C |
MDL Number:
MFCD00068195 CAS Number: 372-29-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...