2-Acetamidothiazole ≥98.0%

Supplier: TCI America

A1472-25G A1472-5G
TCA1472-025GEA 497.54 CAD
TCA1472-025G TCA1472-005G
2-Acetamidothiazole ≥98.0%
2-Acetamidothiazole

CAS Number: 2719-23-5
MDL Number: MFCD00016892
Molecular Formula: C5H6N2OS
Molecular Weight: 142.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR