(S)-Methyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A1466-5G A1466-25G
TCA1466-5GEA 40.5 CAD
TCA1466-5G TCA1466-25G
(S)-Methyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(S)-Methyl-2-aminopropanoate hydrochloride

CAS Number: 2491-20-5
MDL Number: MFCD00063663
Molecular Formula: C4H9NO2
Molecular Weight: 139.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: 8 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride, H-Ala-OMe.HCl

Formula: C₄H₉NO₂·HCl
MW: 139.6 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00063663
CAS Number: 2491-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bbf57abcd6aa5ad6db793322764a824ca13f89d&vid=4bbf57abcd6aa5ad6db793322764a824ca13f89d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR