cis-4-Aminocyclohexanecarboxylic acid ≥98.0%

Supplier: TCI America

A1448-1G A1448-5G A1448-25G
TCA1448-001GEA 39.5 CAD
TCA1448-001G TCA1448-005G TCA1448-25G
cis-4-Aminocyclohexanecarboxylic acid ≥98.0%
cis-4-Aminocyclohexanecarboxylic acid

CAS Number: 3685-23-2
MDL Number: MFCD00191730
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: cis-4-Aminohexahydrobenzoic Acid

Formula: C₇H₁₃NO₂
MDL Number: MFCD00191730
CAS Number: 3685-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5af8e4ddd105349cc2ebb50cf90557a7a8ad6d0&vid=f5af8e4ddd105349cc2ebb50cf90557a7a8ad6d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR