Allyl phenyl sulfide ≥98.0%

Supplier: TCI America

A1442-5G A1442-25G
TCA1442-005GEA 47.4 CAD
TCA1442-005G TCA1442-025G
Allyl phenyl sulfide ≥98.0%
Allyl phenyl sulphide

CAS Number: 5296-64-0
MDL Number: MFCD00014957
Molecular Formula: C9H10S
Molecular Weight: 150.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 98
Specific Gravity (20/20): 1.03

Formula: C₉H₁₀S
MW: 150.24 g/mol
Density: 1.03 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00014957
CAS Number: 5296-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04443896259a5bb5442fc109a1bbc09d8129b471&vid=04443896259a5bb5442fc109a1bbc09d8129b471&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR