(S)-(-)-4-Amino-2-hydroxybutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1438-25G A1438-5G
TCA1438-025GEA 184.86 CAD
TCA1438-025G TCA1438-005G
(S)-(-)-4-Amino-2-hydroxybutyric acid ≥98.0% (by titrimetric analysis)
(S)-(-)-4-Amino-2-hydroxybutyric Acid

CAS Number: 40371-51-5
MDL Number: MFCD00674110
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 203
Specific rotation [a]20/D: -30 deg (C=1, H2O)

Formula: C₄H₉NO₃
Melting Pt: 203 °C
MDL Number: MFCD00674110
CAS Number: 40371-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6285981067e79088add86de962de78c0e284e197&vid=6285981067e79088add86de962de78c0e284e197&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR