beta-Alanine benzyl ester p-toluenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: H-β-Ala-OBzl·p-tosylate, H-β-Ala-Obzl.TosOH, H-β-Ala-OBzl·TosOH

A1423-5G A1423-25G
TCA1423-005GEA 55.3 CAD
TCA1423-005G TCA1423-025G
beta-Alanine benzyl ester p-toluenesulfonate ≥98.0%
beta-Alanine benzyl ester p-toluenesulfonate

CAS Number: 27019-47-2
MDL Number: MFCD00039061
Molecular Formula: C10H13NO2
Molecular Weight: 351.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
MDL Number: MFCD00039061
CAS Number: 27019-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c6d8a352e0e05b4644200a6a97fc22d9a1767ac&vid=7c6d8a352e0e05b4644200a6a97fc22d9a1767ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR