Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate ≥98.0%

Supplier: TCI America

A1354-5G A1354-25G
TCA1354-5GEA 26.86 CAD
TCA1354-5G TCA1354-025G
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate ≥98.0%
Diethyl-5-amino-3-methyl-2,4-thiophenedicarboxylate

CAS Number: 4815-30-9
MDL Number: MFCD00005450
Molecular Formula: C11H15NO4S
Molecular Weight: 257.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: 2-Amino-3,5-bis(ethoxycarbonyl)-4-methylthiophene, 5-Amino-3-methyl-2,4-thiophenedicarboxylic Acid Diethyl Ester

Formula: C₁₁H₁₅NO₄S
Melting Pt: 108 °C
MDL Number: MFCD00005450
CAS Number: 4815-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c7da3b38722930a1d15e98bc6ab9039defc4ee6&vid=8c7da3b38722930a1d15e98bc6ab9039defc4ee6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR