4-Amino-m-toluic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-3-methylbenzoic acid

A1353-25G A1353-5G
TCA1353-25GEA 265.44 CAD
TCA1353-25G TCA1353-5G
4-Amino-m-toluic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-m-toluic acid

CAS Number: 2486-70-6
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 172


Other Product Names / Synonyms: 4-Amino-m-toluic Acid

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00007736
CAS Number: 2486-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7668cc0d2a4941e766977d01775475e5faa27dce&vid=a9792a063c73713731da59a21de1b7a6d2a538e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR