Ethyl-N-hydroxyacetimidate ≥95.0%
Supplier: TCI America
Synonyms:
Ethyl acetohydroximate
A1339-5G
A1339-25G
TCA1339-5GEA
63.2
CAD
TCA1339-5G
TCA1339-25G
Ethyl-N-hydroxyacetimidate ≥95.0%
Ethyl N-hydroxyacetimidate
CAS Number: 10576-12-2
MDL Number: MFCD00002114
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 60
Flash Point (°C): 76
Freezing point (°C): 24
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Acetohydroximic Acid Ethyl Ester, N-Hydroxyacetimidic Acid Ethyl Ester, Ethyl N-Hydroxyacetimidate
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 60 °C Flash Pt: 76 °C |
MDL Number:
MFCD00002114 CAS Number: 10576-12-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...