Methyl-2-acetamidoacrylate ≥97.0%

Supplier: TCI America

A1304-1G A1304-5G
TCA1304-001GEA 109.02 CAD
TCA1304-001G TCA1304-005G
Methyl-2-acetamidoacrylate ≥97.0%
Methyl-2-acetamidoacrylate

CAS Number: 35356-70-8
MDL Number: MFCD00013394
Molecular Formula: C6H9NO3
Molecular Weight: 143.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 104
Melting point (°C): 51
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Acetamidoacrylic Acid Methyl Ester

Formula: C₆H₉NO₃
Boiling Pt: 104 °C
Melting Pt: 51 °C
Flash Pt: 110 °C
MDL Number: MFCD00013394
CAS Number: 35356-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0944bac721643a043883e2285e26d8cac8b37eba&vid=0944bac721643a043883e2285e26d8cac8b37eba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR