(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥99.0% (by GC, titration analysis)

Supplier: TCI America

A1230-1G A1230-5G
TCA1230-1GEA 36.34 CAD
TCA1230-1G TCA1230-5G
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥99.0% (by GC, titration analysis)
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol

CAS Number: 23190-16-1
MDL Number: MFCD00074960
Molecular Formula: C14H15NO
Molecular Weight: 213.28
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Specific rotation [a]20/D: -7 deg (C=0.6, EtOH)

Formula: C₁₄H₁₅NO
Melting Pt: 144 °C
MDL Number: MFCD00074960
CAS Number: 23190-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e57e085ebe0617283f7751d2e8097db68f32bdb0&vid=e57e085ebe0617283f7751d2e8097db68f32bdb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR