Methyl-3-aminopyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Methyl 2-aminopyrazine-3-carboxylate
A1186-5G
A1186-25G
A1186-1G
TCA1186-005GEA
56.88
CAD
TCA1186-005G
TCA1186-25G
TCA1186-001G
Methyl-3-aminopyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Methyl 3-aminopyrazine-2-carboxylate
CAS Number: 16298-03-6
MDL Number: MFCD00010097
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 172
Other Product Names / Synonyms: 3-Aminopyrazine-2-carboxylic Acid Methyl Ester
Formula:
C₆H₇N₃O₂ MW: 153.14 g/mol Melting Pt: 172 °C |
MDL Number:
MFCD00010097 CAS Number: 16298-03-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...