5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride
CAS Number: 72-40-2
MDL Number: MFCD00012704
Molecular Formula: C4H6N4O
Molecular Weight: 162.58
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride
Formula:
C₄H₇ClN₄O MW: 162.58 g/mol |
MDL Number:
MFCD00012704 CAS Number: 72-40-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...