(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1172-25G A1172-5G
TCA1172-25GEA 680.98 CAD
TCA1172-25G TCA1172-005G
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine

CAS Number: 122536-76-9
MDL Number: MFCD00143194
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 78
Specific rotation [a]20/D: -20 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3S)-(-)-3-(Boc-amino)pyrrolidine

Formula: C₉H₁₈N₂O₂
Melting Pt: 78 °C
MDL Number: MFCD00143194
CAS Number: 122536-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66a3e039a2b51ec87d168b045dd5a88102d27609&vid=66a3e039a2b51ec87d168b045dd5a88102d27609&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR