(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate

A1171-5G A1171-25G
TCA1171-005GEA 139.04 CAD
TCA1171-005G TCA1171-25G
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine

CAS Number: 122536-77-0
MDL Number: MFCD00143191
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Specific rotation [a]20/D: 20 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3R)-(+)-3-(Boc-amino)pyrrolidine

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
MDL Number: MFCD00143191
CAS Number: 122536-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35ef8e98809e0448d58740b7a3e1e873a06d9c02&vid=35ef8e98809e0448d58740b7a3e1e873a06d9c02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR