(±)-Dimethyl acetylsuccinate ≥96.0%

Supplier: TCI America

A1143-25G A1143-500G
TCA1143-025GEA 31.6 CAD
TCA1143-025G TCA1143-500G
(±)-Dimethyl acetylsuccinate ≥96.0%
(±)-Dimethyl acetylsuccinate

CAS Number: 10420-33-4
MDL Number: MFCD00008448
Molecular Formula: C8H12O5
Molecular Weight: 188.18
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 200
Flash Point (°C): 110
Freezing point (°C): 30


Other Product Names / Synonyms: Acetylsuccinic Acid Dimethyl Ester

Formula: C₈H₁₂O₅
Boiling Pt: 200 °C
Flash Pt: 110 °C
MDL Number: MFCD00008448
CAS Number: 10420-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8546c3c3d05ec707eb63da1193c0d869c3e16fa1&vid=8546c3c3d05ec707eb63da1193c0d869c3e16fa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR