(-)-O-Acetyl-L-malic anhydride ≥97.0%

Supplier: TCI America

Synonyms: (S)-2-Acetoxysuccinic Anhydride, (S)-(-)-2-Acetoxysuccinic Anhydride

A1101-25G A1101-5G
TCA1101-025GEA 376.83 CAD
TCA1101-025G TCA1101-005G
(-)-O-Acetyl-L-malic anhydride ≥97.0%
(-)-O-Acetyl-L-malic anhydride

CAS Number: 59025-03-5
MDL Number: MFCD00082708
Molecular Formula: C6H6O5
Molecular Weight: 158.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 55
Specific rotation [a]20/D: -25.5 deg (C=5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Acetoxysuccinic Anhydride

Formula: C₆H₆O₅
Melting Pt: 55 °C
MDL Number: MFCD00082708
CAS Number: 59025-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4408d383793de3f798148b974118987a7fffb68&vid=e4408d383793de3f798148b974118987a7fffb68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR