4-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
A1096-25G
A1096-5G
TCA1096-25GEA
472.42
CAD
TCA1096-25G
TCA1096-5G
4-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
4-Aminobenzyl alcohol
CAS Number: 623-04-1
MDL Number: MFCD00014782
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 63
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-(Hydroxymethyl)aniline
Formula:
C₇H₉NO MW: 123.15 Boiling Pt: 171 °C Melting Pt: 63 °C |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...