5-Azacytosine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-1,3,5-triazin-2(1H)-one, 6-Amino-1,3,5-triazin-2(1H)-one, 2-Amino-4-hydroxy-1,3,5-triazine

A1091-25G A1091-5G
TCA1091-025GEA 374.46 CAD
TCA1091-025G TCA1091-005G
5-Azacytosine ≥98.0% (by titrimetric analysis)
5-Azacytosine

CAS Number: 931-86-2
MDL Number: MFCD00006033
Molecular Formula: C3H4N4O
Molecular Weight: 112.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-4-hydroxy-1,3,5-triazine, 4-Amino-1,3,5-triazin-2(1H)-one

Formula: C₃H₄N₄O
MW: 112.09 g/mol
MDL Number: MFCD00149402
CAS Number: 931-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e73a286c9ec4a137cd6d0b31dea2091088256acd&vid=e73a286c9ec4a137cd6d0b31dea2091088256acd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR