1,6-Anhydro-β-D-glucose ≥99.0%

Supplier: TCI America

A1074-1G A1074-5G
TCA1074-001GEA 48.98 CAD
TCA1074-001G TCA1074-005G
1,6-Anhydro-β-D-glucose ≥99.0%
1,6-Anhydro-beta-D-glucose

CAS Number: 498-07-7
MDL Number: MFCD00063248
Molecular Formula: C6H10O5
Molecular Weight: 162.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 183
Specific rotation [a]20/D: -66.5 deg (C=2, H2O)

Formula: C₆H₁₀O₅
Melting Pt: 183 °C
MDL Number: MFCD00063248
CAS Number: 498-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a8e5a854d36d9b8b5a512ff6168477ed17f5791&vid=5a8e5a854d36d9b8b5a512ff6168477ed17f5791&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR