(S)-(+)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1054-5G
A1054-25G
TCA1054-005GEA
352.02
CAD
TCA1054-005G
TCA1054-025G
(S)-(+)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
(S)-(+)-3-Aminopyrrolidine dihydrochloride
CAS Number: 116183-83-6
MDL Number: MFCD00070605
Molecular Formula: C4H10N2
Molecular Weight: 159.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 1.1 deg (C=10, H2O)
Formula:
C₄H₁₂Cl₂N₂ |
MDL Number:
MFCD00070605 CAS Number: 116183-83-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...