1,1'-(Azodicarbonyl)dipiperidine ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Diazene-1,2-diylbis(piperidin-1-ylmethanone), ADDP

A1051-25G A1051-5G
TCA1051-25GEA 489.8 CAD
TCA1051-25G TCA1051-005G
1,1'-(Azodicarbonyl)dipiperidine ≥98.0% (by total nitrogen basis)
1,1'-(Azodicarbonyl)dipiperidine

CAS Number: 10465-81-3
MDL Number: MFCD00010111
Molecular Formula: C12H20N4O2
Molecular Weight: 252.32
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 136


Product Categories: Synthetic Organic Chemistry, Mitsunobu Reaction
Other Product Names / Synonyms: ADDP

Formula: C₁₂H₂₀N₄O₂
MW: 252.32 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00010111
CAS Number: 10465-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf62fc144800823321f9dfe843525c05293c857&vid=3cf62fc144800823321f9dfe843525c05293c857&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR