6-Amino-2-methylquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-methylquinolin-6-amine

A1047-5G A1047-1G
TCA1047-5GEA 1204.91 CAD
TCA1047-5G TCA1047-001G
6-Amino-2-methylquinoline ≥98.0% (by GC, titration analysis)
6-Amino-2-methylquinoline

CAS Number: 65079-19-8
MDL Number: MFCD00052600
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: 6-Aminoquinaldine

Formula: C₁₀H₁₀N₂
MW: 158.20 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00052600
CAS Number: 65079-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=261c166f2ae750e88ad229539032c53ccec2f3fc&vid=261c166f2ae750e88ad229539032c53ccec2f3fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR