L-Azetidine-2-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1043-100MG A1043-1G
TCA1043-100MGEA 79 CAD
TCA1043-100MG TCA1043-1G
L-Azetidine-2-carboxylic acid ≥98.0% (by titrimetric analysis)
L-Azetidine-2-carboxylic acid

[Antagonist of L-Proline]
CAS Number: 2133-34-8
MDL Number: MFCD00005166
Molecular Formula: C4H7NO2
Molecular Weight: 101.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -122 deg (C=4, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: L-Trimethyleneimine-2-carboxylic Acid

Formula: C₄H₇NO₂
MDL Number: MFCD00005166
CAS Number: 2133-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c2712ea8abcfa03df183d4cda5ad827e4467fd0&vid=5c2712ea8abcfa03df183d4cda5ad827e4467fd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR