2'-Methylacetoacetanilide ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylacetoacetanilide, 2-Acetoacetotoluidide

A1020-500G A1020-25G
TCA1020-500GEA 150.1 CAD
TCA1020-500G TCA1020-025G
2'-Methylacetoacetanilide ≥98.0%
2'-Methylacetoacetanilide

CAS Number: 93-68-5
MDL Number: MFCD00008782
Molecular Formula: C11H13NO2
Molecular Weight: 191.23
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 106
Flash Point (°C): 160


Other Product Names / Synonyms: N-Acetoacetyl-2-methylaniline, N-Acetoacetyl-o-toluidine

Formula: C₁₁H₁₃NO₂
Melting Pt: 106 °C
Flash Pt: 160 °C
MDL Number: MFCD00008782
CAS Number: 93-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ecf313bc9c970bd82365dcdd7ae551c11c28595&vid=2ecf313bc9c970bd82365dcdd7ae551c11c28595&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR