4,4'-Anisoin ≥97.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone, Anisoin

A1010-25G A1010-100G
TCA1010-025GEA 96.38 CAD
TCA1010-025G TCA1010-100G
4,4'-Anisoin ≥97.0%
4,4'-Anisoin

CAS Number: 119-52-8
MDL Number: MFCD00008411
Molecular Formula: C16H16O4
Molecular Weight: 272.30
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 112


Other Product Names / Synonyms: 4,4'-Dimethoxybenzoin

Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00008411
CAS Number: 119-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9a063a5d617267eda05bcf2044a5b8abaaf37e6&vid=d9a063a5d617267eda05bcf2044a5b8abaaf37e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR