Methyl-3-aminocrotonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1001-25G
A1001-500G
TCA1001-025GEA
26.86
CAD
TCA1001-025G
TCA1001-500G
Methyl-3-aminocrotonate ≥98.0% (by titrimetric analysis)
Methyl 3-aminocrotonate
CAS Number: 14205-39-1
MDL Number: MFCD00008072
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 83
Flash Point (°C): 91
Other Product Names / Synonyms: 3-Amino-2-butenoic Acid Methyl Ester, 3-Aminocrotonic Acid Methyl Ester, Methyl 3-Amino-2-butenoate
Formula:
C₅H₉NO₂ Melting Pt: 83 °C Flash Pt: 91 °C |
MDL Number:
MFCD00008072 CAS Number: 14205-39-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...