6-Acetoxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0970-5G A0970-1G
TCA0970-005GEA 395 CAD
TCA0970-005G TCA0970-001G
6-Acetoxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
6-Acetoxy-2-naphthoic acid

CAS Number: 17295-26-0
MDL Number: MFCD00059556
Molecular Formula: C13H10O4
Molecular Weight: 230.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₁₃H₁₀O₄
Melting Pt: 228 °C
MDL Number: MFCD00059556
CAS Number: 17295-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57c587c76617ab65cb6097a526cc223cd4770c89&vid=57c587c76617ab65cb6097a526cc223cd4770c89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR