2-(2-Aminothiazol-4-yl)acetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0954-250G A0954-25G
TCA0954-250GEA 479.37 CAD
TCA0954-250G TCA0954-025G
2-(2-Aminothiazol-4-yl)acetic acid ≥98.0% (by HPLC, titration analysis)
2-(2-Aminothiazol-4-yl)acetic acid

CAS Number: 29676-71-9
MDL Number: MFCD00010068
Molecular Formula: C5H6N2O2S
Molecular Weight: 158.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 123
Storage Temperature: 0-10°C

Formula: C₅H₆N₂O₂S
Melting Pt: 123 °C
MDL Number: MFCD00010068
CAS Number: 29676-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0145e6c6893e89ff9cf5e7cd412a472b77032f45&vid=0145e6c6893e89ff9cf5e7cd412a472b77032f45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR