Benzhydrylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0839-250G
A0839-25G
TCA0839-250GEA
444.61
CAD
TCA0839-250G
TCA0839-025G
Benzhydrylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Benzhydrylamine hydrochloride
CAS Number: 5267-34-5
MDL Number: MFCD00013023
Molecular Formula: C13H13N
Molecular Weight: 219.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 293
Other Product Names / Synonyms: alpha-Aminodiphenylmethane Hydrochloride, 1,1-Diphenylmethylamine Hydrochloride
Formula:
C₁₃H₁₄NCl Melting Pt: 293 °C |
MDL Number:
MFCD00013023 CAS Number: 5267-34-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...