Dimethyl adipimidate dihydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Adipimidic Acid Dimethyl Ester Dihydrochloride

A0806-5G A0806-25G
TCA0806-005GEA 109.02 CAD
TCA0806-005G TCA0806-025G
Dimethyl adipimidate dihydrochloride ≥98.0% (by titrimetric analysis)
Dimethyl adipimidate dihydrochloride

[Cross-linking Agent for Protein Research]
CAS Number: 14620-72-5
MDL Number: MFCD00012573
Molecular Formula: C8H16N2O2
Molecular Weight: 245.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 215
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Adipimidic Acid Dimethyl Ester Dihydrochloride

Formula: C₈H₁₆N₂O₂
MW: 245.14 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00012573
CAS Number: 14620-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38803eb5fbc2cab70761bc192d407617a80abf41&vid=38803eb5fbc2cab70761bc192d407617a80abf41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR