6-Aminopenicillanic acid ≥98.0% (by HPLC)

Supplier: TCI America

A0800-5G A0800-25G
TCA0800-005GEA 28.28 CAD
TCA0800-005G TCA0800-025G
6-Aminopenicillanic acid ≥98.0% (by HPLC)
6-Aminopenicillanic acid

CAS Number: 551-16-6
MDL Number: MFCD00005176
Molecular Formula: C8H12N2O3S
Molecular Weight: 216.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 199
Specific rotation [a]20/D: 275 deg (C=1.2, 0.1mol/L HCl)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, 6-APA

Formula: C8H12N2O3S
Melting Pt: 199 °C
MDL Number: MFCD00005176
CAS Number: 551-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cff7566278ed772766e0ca22295c337dae462d10&vid=cff7566278ed772766e0ca22295c337dae462d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR