Sodium-4-aminoazobenzene-4'-sulfonate ≥98.0%

Supplier: TCI America

Synonyms: 4-Aminoazobenzene-4'-sulfonic acid Sodium Salt, 4-Aminoazobenzene-4'-sulphonic acid Sodium Salt

A0748-25G A0748-500G
TCA0748-025GEA 50.56 CAD
TCA0748-025G TCA0748-500G
Sodium-4-aminoazobenzene-4'-sulfonate ≥98.0%
Sodium-4-aminoazobenzene-4'-sulfonate

CAS Number: 2491-71-6
MDL Number: MFCD00035564
Molecular Formula: C12H11N3O3S
Molecular Weight: 299.28
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Aminoazobenzene-4'-sulfonic Acid Sodium Salt

Formula: C₁₂H₁₁N₃O₃S
MDL Number: MFCD00035564
CAS Number: 2491-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8375c48cf69506644d04da2539f7c62432ed0776&vid=8375c48cf69506644d04da2539f7c62432ed0776&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR