Ethyl-1-adamantanecarboxylate ≥98.0%
Supplier: TCI America
A0722-25G
A0722-5G
TCA0722-025GEA
570.38
CAD
TCA0722-025G
TCA0722-005G
Ethyl-1-adamantanecarboxylate ≥98.0%
Ethyl-1-adamantanecarboxylate
CAS Number: 2094-73-7
MDL Number: MFCD00078272
Molecular Formula: C13H20O2
Molecular Weight: 208.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 1-Adamantanecarboxylic Acid Ethyl Ester
Formula:
C₁₃H₂₀O₂ Boiling Pt: 123 °C Density: 1.05 (20 °C) |
MDL Number:
MFCD00078272 CAS Number: 2094-73-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...