Diisobutyl adipate ≥99.0%

Supplier: TCI America

A0706-500ML A0706-25ML
TCA0706-500MLEA 114.16 CAD
TCA0706-500ML TCA0706-025ML
Diisobutyl adipate ≥99.0%
Diisobutyl adipate

CAS Number: 141-04-8
MDL Number: MFCD00053722
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 293
Flash Point (°C): 156
Freezing point (°C): -22
Specific Gravity (20/20): 0.95


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Adipic Acid Diisobutyl Ester

Formula: C₁₄H₂₆O₄
Boiling Pt: 293 °C
Density: 0.95 (20 °C)
Flash Pt: 156 °C
MDL Number: MFCD00053722
CAS Number: 141-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89411dd9c3c5cb43bb26985bdd197c9421e7494a&vid=89411dd9c3c5cb43bb26985bdd197c9421e7494a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR