2-Acetamido-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(5-Nitro-2-thiazolyl)acetamide, N-(5-Nitrothiazol-2-yl)acetamide

A0689-5G A0689-1G
TCA0689-005GEA 315.68 CAD
TCA0689-005G TCA0689-1G
2-Acetamido-5-nitrothiazole ≥98.0% (by HPLC, titration analysis)
2-Acetamidot-5-nitrohiazole

CAS Number: 140-40-9 MDL Number: MFCD00022438 Molecular Formula: C5H5N3O3S Molecular Weight: 187.17 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal

Formula: C₅H₅N₃O₃S
MW: 187.18 g/mol
MDL Number: MFCD00022438
CAS Number: 140-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f927686d15c07f2abf67404adb5bfb654cb93645&vid=f927686d15c07f2abf67404adb5bfb654cb93645&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR