Ammeline ≥95.0%
Supplier: TCI America
Synonyms:
Cyanurodiamide, 4,6-Diamino-1,3,5-triazin-2-ol, 2,4-Diamino-6-hydroxy-1,3,5-triazine, 3,4,5,6-Tetrahydro-4,6-diimino-1,3,5-triazin-2(1H)-one
CAS Number: 645-92-1
MDL Number: MFCD00023186
Molecular Formula: C3H5N5O
Molecular Weight: 127.11
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Other Product Names / Synonyms: Cyanurodiamide, 2,4-Diamino-6-hydroxy-1,3,5-triazine, 4,6-Diamino-1,3,5-triazin-2-ol, 3,4,5,6-Tetrahydro-4,6-diimino-1,3,5-triazin-2(1H)-one
Formula:
C₃H₅N₅O MW: 127.11 g/mol |
MDL Number:
MFCD00023186 CAS Number: 645-92-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...