m-Tolyl acetate ≥99.0%
Supplier: TCI America
A0659-25G
A0659-500G
TCA0659-025GEA
82.16
CAD
TCA0659-025G
TCA0659-500G
m-Tolyl acetate ≥99.0%
m-Tolyl acetate
CAS Number: 122-46-3
MDL Number: MFCD00041910
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Melting point (°C): 12
Flash Point (°C): 95
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Acetic Acid m-Cresyl Ester, Acetic Acid m-Tolyl Ester, m-Cresyl Acetate
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 212 °C Melting Pt: 12 °C Density: 1.05 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00041910 CAS Number: 122-46-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...