N-Acetylneuraminic acid hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0639-5G
A0639-1G
A0639-100MG
TCA0639-5GEA
254.02
CAD
TCA0639-5G
TCA0639-001G
TCA0639-100MG
N-Acetylneuraminic acid hydrate ≥98.0% (by titrimetric analysis)
N-Acetylneuraminic acid hydrate
CAS Number: 131-48-6
MDL Number: MFCD00006620
Molecular Formula: C11H19NO9
Molecular Weight: 309.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 186
Specific rotation [a]20/D: -32 deg (C=1, H2O)
Storage Temperature: <0°C
Product Categories: Bioscience, Sugar Acids, Sialic Acids
Other Product Names / Synonyms: NANA, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic Acid, Neu5Ac
Formula:
C₁₁H₁₉NO₉ Melting Pt: 186 °C |
MDL Number:
MFCD00006620 CAS Number: 131-48-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...