9-Aminofluorene hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0622-5G
A0622-25G
TCA0622-005GEA
218.2
CAD
TCA0622-005G
TCA0622-025G
9-Aminofluorene hydrochloride ≥98.0% (by HPLC, titration analysis)
9-Aminofluorene hydrochloride
CAS Number: 5978-75-6
MDL Number: MFCD00012536
Molecular Formula: C13H11N
Molecular Weight: 217.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 257
Other Product Names / Synonyms: 9-Fluorenamine Hydrochloride
Formula:
C₁₃H₁₁N Melting Pt: 257 °C |
MDL Number:
MFCD00012536 CAS Number: 5978-75-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...