Azure II

Supplier: TCI America

A0574-25G A0574-1G
TCA0574-025GEA 61.62 CAD
TCA0574-025G TCA0574-001G
Azure II
Azure II

CAS Number: 37247-10-2
MDL Number: MFCD00080716
Form: Crystal
Color: Yellow Green
Melting point (°C): 185
Lambda max.: 657 nm (H2O)


Other Product Names / Synonyms: Methylene Azure II

Formula: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
Melting Pt: 185 °C
MDL Number: MFCD00080716
CAS Number: 37247-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR